Proteintech
Manufacturer Code:171251AP150UL
Catalog # 50-173-1396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CSNK1A1L Fusion Protein Ag10814 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: casein kinase 1 alpha 1 like CK1 CKI alpha like CSNK1A1L RP11 532O21.2; casein kinase 1, alpha 1-like; casein kinase I alpha S-like; Casein kinase I isoform alpha-like; CK1; CKI-alpha-like
Gene Aliases: CK1; CSNK1A1L
UniProt ID: (Human) Q8N752
Entrez Gene ID: (Human) 122011
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.