Proteintech
Manufacturer Code:551921AP150UL
Catalog # 50-554-364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: casein kinase 1 alpha 1 Casein kinase I isoform alpha CK1 CKI alpha CSNK1A1 HLCDGP1 PRO2975; casein kinase 1, alpha 1; Casein kinase I isoform alpha; casein kinase I-alpha; CK1; CKI-alpha; clock regulator kinase; down-regulated in lung cancer; epididymis secretory sperm binding protein Li 77p
Gene Aliases: 2610208K14Rik; 4632404G05Rik; 5430427P18Rik; CK1; CK1a; CKIa; Csnk1a; CSNK1A1; HEL-S-77p; HLCDGP1; PRO2975
UniProt ID: (Human) P48729, (Rat) P97633
Entrez Gene ID: (Human) 1452, (Rat) 113927, (Mouse) 93687
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.