Novus Biologicals
Manufacturer Code:NBP288633
Catalog # NB036255
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of CSN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COP9 signalosome complex subunit 1; COP9 signalosome complex subunit 1 COPS1JAB1-containing signalosome subunit 1 CSN1MGC71287 G protein pathway suppressor 1SGN1 GPS-1 Protein MFH Signalosome subunit 1; G protein pathway suppressor 1; GPS-1; JAB1-containing signalosome subunit 1; Protein MFH; SGN1
Gene Aliases: COPS1; CSN1; GPS1; SGN1
UniProt ID: (Human) Q13098
Entrez Gene ID: (Human) 2873
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.