Novus Biologicals
Manufacturer Code:NBP188086
Catalog # NBP188086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:PCVEAVGERGGPQNPDSRARLDQSDEDFKPRIVPYYRDPNKPYKKVLRTRYIQTELGSRERLLVAVLTSRATLST |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chondroitin glucuronyltransferase; Chondroitin glucuronyltransferase chondroitin polymerizing factor 2KIAA1402chPF-2 Chondroitin synthase 3 chondroitin synthase-3 ChPF-2 CHSY3 ChSy-3chondroitin sulfate glucuronyltransferase CSGlcAT CSGlcA-T EC 2.4.1.226 N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase; Chondroitin polymerizing factor 2; Chondroitin sulfate glucuronyltransferase; Chondroitin synthase 3; chondroitin synthase-3; ChPF-2; ChSy-3; CSGlcA-T; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase
Gene Aliases: chPF-2; CHPF2; ChSy-3; CHSY3; CSGLCA-T; CSGLCAT; KIAA1402; UNQ299/PRO339
UniProt ID: (Human) Q9P2E5
Entrez Gene ID: (Human) 54480
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.