Invitrogen
Manufacturer Code:PA524847
Catalog # PIPA524847
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 192-220 amino acids from the central region of human CSGALNACT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta1,4 N-acetylgalactosaminyltransferase; Beta4GalNAcT-1; Chondroitin beta-1,4-N-acetylgalactosaminyltransferase 1; chondroitin beta1,4 N-acetylgalactosaminyltransferase; Chondroitin sulfate N-acetylgalactosaminyltransferase 1; CsGalNAcT-1; CSGalNAcT-1 ChGn beta4GalNAcT beta4GalNAcT-1 chondroitin beta-14-N-acetylgalactosaminyltransferase 1 chondroitin beta14 N-acetylgalactosaminyltransferase; glucuronylgalactosylproteoglycan 4-beta-N- acetylgalactosaminyltransferase
Gene Aliases: 4732435N03Rik; beta4GalNAcT; CHGN; CSGalNAcT-1; CSGALNACT1; GALNACT1; UNQ656/PRO1287
UniProt ID: (Human) Q8TDX6, (Mouse) Q8BJQ9
Entrez Gene ID: (Human) 55790, (Mouse) 234356
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.