OriGene
Manufacturer Code:TA807874
Catalog # 50-168-1820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A6 |
Immunogen | Full length human recombit protein of human CSF2RB produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: beta common cytokine receptor; CD131; CD131 CDw131 IL3RB IL5RB SMDP5 betaGMR; CDw131; colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage); colony-stimulating factor-2 receptor, beta, low-affinity; Cytokine receptor common subunit beta; GM-CSF/IL-3/IL-5 receptor common beta subunit; GM-CSF/IL-3/IL-5 receptor common beta-chain; interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3 receptor, beta (high affinity)
Gene Aliases: CD131; CDw131; CSF2RB; IL3RB; IL5RB; SMDP5
UniProt ID: (Human) P32927
Entrez Gene ID: (Human) 1439
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.