Invitrogen
Manufacturer Code:PA548082
Catalog # PIPA548082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse IL-3 R beta His23-Trp440 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AIC2A BetaIl3 Bil3 Csfgmrb IL3R IL3RB Il3rb2; Colony-stimulating factor 2 receptor subunit beta-2; IL-3 receptor class 2 subunit beta; IL-3R class 2 subunit beta; Interleukin-3 receptor class 2 subunit beta; Interleukin-3 receptor class II beta chain
Gene Aliases: Ai2ca; AIC2A; BetaIl3; Bil3; Csf2rb2; Csfgmrb; Il3r; Il3rb; Il3rb2
UniProt ID: (Mouse) P26954
Entrez Gene ID: (Mouse) 12984
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.