OriGene
Manufacturer Code:CF808055
Catalog # 50-166-8619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10G3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 18-144 of human CSF2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: colony stimulating factor 2 (granulocyte-macrophage); Colony-stimulating factor; CSF; CSF GMCSF; GM-CSF; granulocyte macrophage-colony stimulating factor; Granulocyte-macrophage colony-stimulating factor; Molgramostin; Sargramostim
Gene Aliases: CSF2; GMCSF
UniProt ID: (Human) P04141
Entrez Gene ID: (Human) 1437
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.