Abnova
Manufacturer Code:MAB6297
Catalog # 89-112-808
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 21 |
Immunogen | Recombinant protein corresponding to human CSF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot after reconstitution to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant CSF1.
For Research Use Only
Protein Aliases: colony stimulating factor 1 (macrophage); CSF-1; Lanimostim; macrophage colony stimulating factor 1; Macrophage colony-stimulating factor 1; MCSF; MGC31930; Processed macrophage colony-stimulating factor 1
Gene Aliases: CSF-1; CSF1; MCSF
UniProt ID: (Human) P09603
Entrez Gene ID: (Human) 1435
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.