Invitrogen
Manufacturer Code:PA522394
Catalog # PIPA522394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 198 and 433 of CSDE1 (Uniprot IDNo.O75534) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cold shock domain containing E1, RNA binding; cold shock domain containing E1, RNA-binding; Cold shock domain-containing protein E1; D1S155E RP5-1000E10.3 UNR N-ras upstream gene protein NRAS-related cold shock domain-containing protein E1 upstream of NRAS; N-ras upstream gene protein; NRAS-related; Protein UNR; upstream of NRAS
Gene Aliases: CSDE1; D1S155E; KIAA0885; NRU; UNR
UniProt ID: (Human) O75534
Entrez Gene ID: (Human) 7812
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.