Invitrogen
Manufacturer Code:PA585198
Catalog # PIPA585198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human CSB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARMD5 antibody CKN2 antibody COFS antibody COFS1 antibody CSB antibody RAD26 antibody ERCC6 antibody Cockayne syndrome B protein antibody ATP-dependent helicase ERCC6 antibody cockayne syndrome protein CSB antibody Rad26 homolog antibody Cockayne syndrome group B protein antibody DNA excision repair protein ERCC-6 antibody excision repair cross-complementing rodent repair deficiency complementation group 6 antibody; ATP-dependent helicase ERCC6; Chimeric CSB-PGBD3 protein; Chimeric ERCC6-PGBD3 protein; Cockayne syndrome group B protein; Cockayne syndrome protein CSB; CS group B correcting; DNA excision repair protein ERCC-6; ERCC6-PGBD3 fusion protein (isoform 1); excision repair cross-complementation group 6; excision repair cross-complementing rodent repair deficiency, complementation group 6
Gene Aliases: 4732403I04; ARMD5; C130058G22Rik; CKN2; COFS; COFS1; CSB; ERCC6; POF11; RAD26; UVSS1
UniProt ID: (Human) P0DP91, (Mouse) F8VPZ5
Entrez Gene ID: (Human) 2074, (Rat) 306274, (Mouse) 319955
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.