Novus Biologicals
Manufacturer Code:NBP17045920UL
Catalog # NBP17045920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to C1ORF96 The peptide sequence was selected from the N terminal of C1ORF96. Peptide sequence LGRRLLEQAHAPWLWDDWGPAGSSEDSASSESSGAGGPAPRCAPPSPPPP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf96 centriole cilia and spindle-associated protein chromosome 1 open reading frame 96 RP4-803J11.3; centriole and spindle-associated protein; Centriole, cilia and spindle-associated protein
Gene Aliases: C1orf96; CCSAP; CSAP
UniProt ID: (Human) Q6IQ19
Entrez Gene ID: (Human) 126731
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.