Novus Biologicals
Manufacturer Code:NBP2298370.025MG
Catalog # NB019484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | P8G6 |
Immunogen | IL-1beta activated HUVEC (human). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Aspartate 1-decarboxylase; CSDMGC119355 cysteine sulfinic acid decarboxylase Cysteine-sulfinate decarboxylase EC 4.1.1 EC 4.1.1.29 FLJ44987 FLJ45500 MGC119354 MGC119357 PCAP P-selectin cytoplasmic tail-associated protein Sulfinoalanine decarboxylase; Cysteine sulfinic acid decarboxylase; cysteine sulfinic acid decarboxylase-related protein; Cysteine-sulfinate decarboxylase; P-selectin cytoplasmic tail-associated protein; Sulfinoalanine decarboxylase
Gene Aliases: CSAD; CSD; PCAP
UniProt ID: (Human) Q9Y600
Entrez Gene ID: (Human) 51380
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.