Abnova
Manufacturer Code:H00051380M02A
Catalog # 89-106-895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C11 |
Immunogen | CSAD (NP_0570731 a.a. &bsim 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant CSAD.
For Research Use Only
Protein Aliases: Aspartate 1-decarboxylase; CSD; Cysteine sulfinic acid decarboxylase; cysteine sulfinic acid decarboxylase-related protein; Cysteine-sulfinate decarboxylase; FLJ44987; FLJ45500; MGC119354; MGC119355; MGC119357; P-selectin cytoplasmic tail-associated protein; PCAP; Sulfinoalanine decarboxylase
Gene Aliases: CSAD; CSD; PCAP
UniProt ID: (Human) Q9Y600
Entrez Gene ID: (Human) 51380
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.