Novus Biologicals
Manufacturer Code:NBP271522AF647
Catalog # N271522A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F9 |
Immunogen | Full length recombinant protein of human CRYZL1 (NP_665857) produced in HEK293T cells. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: crystallin zeta (quinone reductase)-like 1 human quinone oxidoreductase homolog-1104P11QOH-1quinone oxidoreductase-like protein 1 Protein 4P11 Quinone oxidoreductase homolog 1 quinone reductase-like 1 Zeta-crystallin homolog; crystallin, zeta (quinone reductase)-like 1; Protein 4P11; QOH-1; Quinone oxidoreductase homolog 1; Quinone oxidoreductase-like protein 1; quinone reductase-like 1; Zeta-crystallin homolog
Gene Aliases: 4P11; CRYZL1; QOH-1
UniProt ID: (Human) O95825
Entrez Gene ID: (Human) 9946
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.