Invitrogen
Manufacturer Code:PA542798
Catalog # PIPA542798
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human CRYZL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: crystallin zeta (quinone reductase)-like 1 4P11 QOH-1; crystallin, zeta (quinone reductase)-like 1; crystallin, zeta like 1; Protein 4P11; QOH-1; Quinone oxidoreductase homolog 1; Quinone oxidoreductase-like protein 1; quinone reductase-like 1; Zeta-crystallin homolog
Gene Aliases: 2210407J23Rik; 2410006O11Rik; 4P11; CRYZL1; dZ63M10.5; QOH-1; zgc:92225
UniProt ID: (Bovine) Q59A28, (Human) O95825, (Mouse) Q921W4
Entrez Gene ID: (Dog) 478408, (Pig) 100049689, (Rabbit) 100358042, (Bovine) 506426, (Human) 9946, (Horse) 100052191, (Mouse) 66609, (Guinea pig) 100728850, (Rat) 288256, (Zebrafish) 436906
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.