Invitrogen
Manufacturer Code:PA588812
Catalog # PIPA588812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-329 of human CRYZ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: crystallin, zeta (quinone reductase); NADPH:quinone reductase; Quinone oxidoreductase; quinone reductase; Zeta-crystallin
Gene Aliases: CRYZ; Sez9
UniProt ID: (Human) Q08257, (Rat) Q6AYT0, (Mouse) P47199
Entrez Gene ID: (Human) 1429, (Rat) 362061, (Mouse) 12972
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.