Novus Biologicals
Manufacturer Code:NBP216016
Catalog # NBP216016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human CRYZ. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: crystallin zeta (quinone reductase) DKFZp779E0834 EC 1.6.5.5 FLJ41475 NADPH:quinone reductase quinone oxidoreductase Zeta-crystallin; crystallin, zeta (quinone reductase); NADPH:quinone reductase; Quinone oxidoreductase; Zeta-crystallin
Gene Aliases: CRYZ
UniProt ID: (Human) Q08257
Entrez Gene ID: (Human) 1429
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.