Abnova
Manufacturer Code:H00001420M01C
Catalog # 89-021-177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7C4 |
Immunogen | CRYGC (NP_066269 75 a.a. ~ 174 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant CRYGC.
For Research Use Only
Protein Aliases: CCL; CRYG3; crystallin, gamma C; Gamma-C-crystallin; Gamma-crystallin 2-1; Gamma-crystallin 3; Gamma-crystallin C
Gene Aliases: CCL; CRYG3; CRYGC; CTRCT2
UniProt ID: (Human) P07315
Entrez Gene ID: (Human) 1420
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.