Invitrogen
Manufacturer Code:PA543812
Catalog # PIPA543812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CRYGC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Crystallin gamma C CCL CRYG3; crystallin, gamma C; crystallin, gamma polypeptide 3; Gamma-C-crystallin; Gamma-crystallin 2-1; Gamma-crystallin 3; Gamma-crystallin C; Gamma-crystallin IIIA; gammaC-crystallin
Gene Aliases: CCL; Cryg; CRYG3; CRYGC; CTRCT2; Len
UniProt ID: (Dog) A3RLE2, (Bovine) Q28088, (Human) P07315, (Rat) P02529
Entrez Gene ID: (Dog) 100126770, (Bovine) 281722, (Human) 1420, (Rabbit) 100354942, (Rat) 24277
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.