Invitrogen
Manufacturer Code:PA5109596
Catalog # PIPA5109596
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-174 of human CRYGC (NP_066269.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOS_2012 CCL Cryg CRYG3 Cryg-5 Crygc CTRCT2 Gammab-cry Gamma-C-crystallin Gamma-crystallin 2-1 Gamma-crystallin 3 Gamma-crystallin C Gamma-crystallin IIIA Len; crystallin, gamma C; Gamma-C-crystallin; Gamma-crystallin 2-1; Gamma-crystallin 3; Gamma-crystallin C
Gene Aliases: CCL; CRYG3; CRYGC; CTRCT2
UniProt ID: (Human) P07315
Entrez Gene ID: (Human) 1420
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.