Proteintech
Manufacturer Code:179311AP150UL
Catalog # 50-173-1883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CRYGC Fusion Protein Ag12362 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCL CRYG3 CRYGC crystallin gamma C Gamma C crystallin Gamma crystallin 2 1 Gamma crystallin 3 Gamma crystallin C; crystallin, gamma C; crystallin, gamma polypeptide 3; Gamma-C-crystallin; Gamma-crystallin 2-1; Gamma-crystallin 3; Gamma-crystallin C; gammaC-crystallin
Gene Aliases: CCL; Cryg; Cryg-5; CRYG3; CRYGC; CTRCT2; Gammab-cry; Len
UniProt ID: (Human) P07315, (Rat) P02529, (Mouse) Q61597
Entrez Gene ID: (Human) 1420, (Rat) 24277, (Mouse) 12966
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.