Invitrogen
Manufacturer Code:PA577837
Catalog # PIPA577837
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human alpha B crystallin conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AACRYA Alpha crystallin B chain CRYA2 CRYAB CTPP2 HSPB5 NY REN 27 antigen; alpha B-crystallin; alpha crystallin B; Alpha(B)-crystallin; Alpha-crystallin B chain; crystallin, alpha 2; crystallin, alpha B; crystallin, alpha polypeptide 2; epididymis secretory protein Li 101; Heat shock protein beta-5; heat shock protein CryAB; heat shock protein HspB5; heat-shock 20 kD like-protein; HspB5; HspB5 protein; P23; Renal carcinoma antigen NY-REN-27; Rosenthal fiber component
Gene Aliases: AACRYA; alpha(B)-crystallin; CMD1II; Crya-2; CRYA2; CRYAB; CTPP2; CTRCT16; HEL-S-101; HSPB5; MFM2; P23
UniProt ID: (Chicken) Q05713, (Human) P02511, (Bovine) P02510, (Rat) P23928, (Mouse) P23927
Entrez Gene ID: (Chicken) 396089, (Human) 1410, (Bovine) 281719, (Rat) 25420, (Mouse) 12955
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.