Novus Biologicals
Manufacturer Code:NBP270449AF647
Catalog # N270449A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H5 |
Immunogen | Full length recombinant protein of human CRY2 (NP_0066940) produced in HEK293T cells. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: cryptochrome 2 (photolyase-like); cryptochrome 2 (photolyase-like) cryptochrome-2 FLJ10332 growth-inhibiting protein 37 HCRY2 KIAA0658 PHLL2; Cryptochrome-2; growth-inhibiting protein 37
Gene Aliases: CRY2; HCRY2; KIAA0658; PHLL2
UniProt ID: (Human) Q49AN0
Entrez Gene ID: (Human) 1408
Molecular Function:
DNA binding protein
DNA photolyase
lyase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.