Invitrogen
Manufacturer Code:MA537755
Catalog # PIMA537755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1819CT334.92.36.9 |
Immunogen | recombinat protein of human Cry2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AV006279 CRY2 cryptochrome 2 (photolyase-like) cryptochrome circadian clock 2 cryptochrome-2 D130054K12Rik growth-inhibiting protein 37 HCRY2 KIAA0658 PHLL2; cryptochrome 2 (photolyase-like); Cryptochrome-2; growth-inhibiting protein 37
Gene Aliases: AV006279; CRY2; D130054K12Rik; HCRY2; KIAA0658; PHLL2
UniProt ID: (Human) Q49AN0, (Mouse) Q9R194
Entrez Gene ID: (Human) 1408, (Mouse) 12953
Molecular Function:
DNA binding protein
DNA photolyase
lyase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.