Invitrogen
Manufacturer Code:PA589349
Catalog # PIPA589349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-255 of human CRY1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cryptochrome 1 (photolyase-like); Cryptochrome-1
Gene Aliases: AU020726; AU021000; CRY1; PHLL1
UniProt ID: (Human) Q16526, (Rat) Q32Q86, (Mouse) P97784
Entrez Gene ID: (Human) 1407, (Rat) 299691, (Mouse) 12952
Molecular Function: DNA binding protein DNA photolyase lyase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.