Novus Biologicals
Manufacturer Code:NBP30375120UL
Catalog # NB093679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 507-586 of human CRY1 (NP_004066.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cryptochrome 1 (photolyase-like); cryptochrome 1 (photolyase-like) PHLL1cryptochrome-1 photolyase-like cryptochrome 1; Cryptochrome-1
Gene Aliases: CRY1; PHLL1
UniProt ID: (Human) Q16526
Entrez Gene ID: (Human) 1407
Molecular Function: DNA binding protein DNA photolyase lyase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.