Invitrogen
Manufacturer Code:PA520332
Catalog # PIPA520332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide from near the amino terminus of human CRTH2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD294; CD294 CRTH2 DL1R DP2 GPR44 G protein-coupled receptor 44 G Protein-Coupled Rec. 44; chemoattractant receptor homologous molecule expressed on T helper type 2 cells; Chemoattractant receptor-homologous molecule expressed on Th2 cells; G protein-coupled receptor 44; G-protein coupled receptor 44; PGD receptor; PGD2 receptor; prostaglandin D receptor; Prostaglandin D2 receptor; prostaglandin D2 receptor (DP); Prostaglandin D2 receptor 2; prostaglandin D2 receptor CRTH2; Prostanoid DP receptor; putative G-protein coupled receptor 44
Gene Aliases: AS1; ASRT1; CD294; CRTH2; DL1R; DP; DP1; DP2; GPR44; Grp45; PGD; PTGDR; PTGDR1; PTGDR2; RGD1565108
UniProt ID: (Human) Q13258, (Human) Q9Y5Y4, (Rat) Q9R261, (Mouse) P70263, (Mouse) Q9Z2J6, (Rat) Q6XKD3
Entrez Gene ID: (Human) 5729, (Human) 11251, (Rat) 498475, (Mouse) 19214, (Mouse) 14764, (Rat) 309212
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.