Novus Biologicals
Manufacturer Code:NB10093564SS
Catalog # NB017917
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 329-348 [DDSELGGAGSSRRRRTSSTA] of the CRTH2 (GPR44) protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD294; CD294 CD294 antigen Chemoattractant receptor-homologous molecule expressed on Th2 cells CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells DL1R DP2 G protein-coupled receptor 44 putative G-protein coupled receptor 44; chemoattractant receptor homologous molecule expressed on T helper type 2 cells; Chemoattractant receptor-homologous molecule expressed on Th2 cells; G protein-coupled receptor 44; G-protein coupled receptor 44; Prostaglandin D2 receptor 2; putative G-protein coupled receptor 44
Gene Aliases: CD294; CRTH2; DL1R; DP2; GPR44; PTGDR2
UniProt ID: (Human) Q9Y5Y4
Entrez Gene ID: (Human) 11251
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.