Proteintech
Manufacturer Code:124971AP150UL
Catalog # 50-172-8025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CRTC2TORC2 Fusion Protein Ag3167 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CREB-regulated transcription coactivator 2; CRTC2 RP11 422P24.6 TORC 2 TORC2 Transducer of CREB protein 2; TORC-2; transducer of CREB protein 2; transducer of regulated cAMP response element-binding protein (CREB) 2; Transducer of regulated cAMP response element-binding protein 2
Gene Aliases: 4632407F12Rik; CRTC2; mTORC2; RGD1308903; TORC-2; TORC2
UniProt ID: (Human) Q53ET0, (Rat) Q3LRZ1, (Mouse) Q3U182
Entrez Gene ID: (Human) 200186, (Rat) 310615, (Mouse) 74343
Molecular Function: CREB transcription factor basic leucine zipper transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.