Invitrogen
Manufacturer Code:PA560431
Catalog # PIPA560431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CRTAP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cartilage-associated protein; CASP LEPREL3 OI7 cartilage-associated protein leprecan-like 3; leprecan-like 3; prolyl 3-hydroxylase family member 5 (non-enzymatic)
Gene Aliases: CASP; CRTAP; LEPREL3; OI7; P3H5
UniProt ID: (Human) O75718
Entrez Gene ID: (Human) 10491
Molecular Function: extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.