Invitrogen
Manufacturer Code:PA542913
Catalog # PIPA542913
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human CRTAP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cartilage-associated protein; cartilage-associated-like protein; leprecan-like 3; prolyl 3-hydroxylase family member 5 (non-enzymatic)
Gene Aliases: 5730529N23Rik; CASP; CRTAP; LEPREL3; OI7; P3H5; RGD1565180; wu:fb47h01; zgc:85621
UniProt ID: (Human) O75718, (Mouse) Q9CYD3
Entrez Gene ID: (Rabbit) 100339498, (Bovine) 540924, (Horse) 100056249, (Human) 10491, (Dog) 485577, (Rat) 363158, (Guinea pig) 100721078, (Mouse) 56693, (Zebrafish) 406626
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.