Proteintech
Manufacturer Code:241751AP150UL
Catalog # 50-173-4254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CRP Fusion Protein Ag21466 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C reactive protein CRP PTX1; C-reactive protein; C-reactive protein(1-205); C-reactive protein, petaxin related; pentraxin 1
Gene Aliases: AI255847; CRP; PTX1
UniProt ID: (Human) P02741, (Mouse) P14847
Entrez Gene ID: (Human) 1401, (Mouse) 12944
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.