Invitrogen
Manufacturer Code:PA543239
Catalog # PIPA543239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human CROT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carnitine O-octanoyltransferase COT; COT; peroxisomal carnitine acyltransferase; Peroxisomal carnitine O-octanoyltransferase
Gene Aliases: 1200003H03Rik; COT; CROT; zgc:110643
UniProt ID: (Bovine) O19094, (Human) Q9UKG9, (Mouse) Q9DC50, (Rat) P11466
Entrez Gene ID: (Rabbit) 100355549, (Bovine) 281092, (Pig) 100521142, (Horse) 100050111, (Dog) 482283, (Human) 54677, (Mouse) 74114, (Rat) 83842, (Zebrafish) 553204
Molecular Function:
acetyltransferase
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.