Invitrogen
Manufacturer Code:PA537981
Catalog # PIPA537981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence NYFKTESEGHCPTH from the internal region of rat Crot (aa178-199) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: carnitine O-octanoyltransferase COT peroxisomal carnitine O-octanoyltransferase; COT; peroxisomal carnitine acyltransferase; Peroxisomal carnitine O-octanoyltransferase
Gene Aliases: 1200003H03Rik; COT; CROT
UniProt ID: (Human) Q9UKG9, (Mouse) Q9DC50
Entrez Gene ID: (Human) 54677, (Mouse) 74114
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.