Invitrogen
Manufacturer Code:PA5104118
Catalog # PIPA5104118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CROCC corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ciliary rootlet coiled-coil protein; Ciliary rootlet coiled-coil protein ciliary rootlet coiled-coil rootletin CROCC KIAA0445 ROLT Rootletin rootletin ciliary rootlet protein Tax1-binding protein 2; CROCC; Rootletin; rootletin, ciliary rootlet protein; Tax1-binding protein 2
Gene Aliases: CROCC; KIAA0445; ROLT
UniProt ID: (Human) Q5TZA2, (Mouse) Q8R0B9
Entrez Gene ID: (Human) 9696, (Mouse) 230872
Molecular Function:
enzyme modulator
kinase modulator
viral protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.