Proteintech
Manufacturer Code:189691AP150UL
Catalog # 50-608-308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CRMP4 Fusion Protein Ag5299 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Collapsin response mediator protein 4; collapsin response mediator protein 4 long; CRMP 4 CRMP4 dihydropyrimidinase like 3 DPYSL3 DRP 3 DRP3 LCRMP ULIP ULIP 1 ULIP1 Unc 33 like phosphoprotein 1; CRMP-4; dihydropyrimidinase-like 3; Dihydropyrimidinase-related protein 3; DRP-3; testicular secretory protein Li 7; TOAD-64/Ulip/CRMP; TOAD-64/Ulip/CRMP protein 4b; TUC-4b; ULIP-1; Unc-33-like phosphoprotein 1
Gene Aliases: CRMP-4; CRMP4; DPYSL3; DRP-3; DRP3; LCRMP; TUC-4b; TUC4; ULIP; ULIP-1; ULIP1
UniProt ID: (Human) Q14195, (Mouse) Q62188, (Rat) Q62952
Entrez Gene ID: (Human) 1809, (Mouse) 22240, (Rat) 25418
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.