Novus Biologicals
Manufacturer Code:NBP30298920UL
Catalog # NB095297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 400-500 of human CRMP2 (NP_001377.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Collapsin response mediator protein 2; Collapsin response mediator protein 2 CRMP-2 CRMP2N2A3 DHPRP2 dihydropyrimidinase-like 2 dihydropyrimidinase-like 2 long form dihydropyrimidinase-related protein 2 DRP2collapsin response mediator protein hCRMP-2 DRP-2ULIP-2 ULIP2 Unc-33-like phosphoprotein 2; collapsin response mediator protein hCRMP-2; CRMP-2; dihydropyrimidinase-like 2; Dihydropyrimidinase-related protein 2; DRP-2; N2A3; ULIP-2; Unc-33-like phosphoprotein 2
Gene Aliases: CRMP-2; CRMP2; DHPRP2; DPYSL2; DRP-2; DRP2; N2A3; ULIP-2; ULIP2
UniProt ID: (Human) Q16555
Entrez Gene ID: (Human) 1808
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.