Invitrogen
Manufacturer Code:PA3120
Catalog # APSA20
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues Y(499) D G P V F D L T T T P K(511) of TUC-4a (residues 612-624 of TUC-4b). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Collapsin response mediator protein 2; CRMP-2; Dihydropyrimidinase-related protein 2; DRP-2; TOAD-64; Turned On After Division 64 DRP2 CRMP-2 DPYL2 Dihydropyrimidinase-related protein 2 Collapsin response mediator protein 2; Turned on after division 64 kDa protein; turned on after division, 64 kDa protein; ULIP-2; Unc-33-like phosphoprotein 2
Gene Aliases: AI851130; Crmp2; Dpysl2; DRP2; Musunc33; TOAD-64; Ulip2
UniProt ID: (Rat) P47942, (Mouse) O08553
Entrez Gene ID: (Rat) 25416, (Mouse) 12934
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.