Abnova
Manufacturer Code:PAB9845
Catalog # 89-134-450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human CRMP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CRMP1.
For Research Use Only
Protein Aliases: Collapsin response mediator protein 1; CRMP-1; dihydropyrimidinase-like 1; Dihydropyrimidinase-related protein 1; DPYSL1 DRP-1 DRP1; DRP-1; Inactive dihydropyrimidinase; ULIP-3; Unc-33-like phosphoprotein 3
Gene Aliases: CRMP-1; CRMP1; DPYSL1; DRP-1; DRP1; ULIP-3; ULIP3
UniProt ID: (Human) Q14194
Entrez Gene ID: (Human) 1400
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.