Invitrogen
Manufacturer Code:MA527879
Catalog # PIMA527879
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5G3 |
Immunogen | E.coli-derived human CRM1 recombit protein (Position: N966-D1071). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: chromosome region maintenance 1 homolog; Chromosome region maintenance 1 protein homolog; Exp1; exportin 1 (CRM1 homolog, yeast); exportin 1 (CRM1, yeast, homolog); exportin 1, CRM1 homolog; Exportin-1; exportin-1 (required for chromosome region maintenance); Exportin-1 Exp1 Chromosome region maintece 1 protein homolog XPO1 CRM1
Gene Aliases: AA420417; CRM1; emb; Exp1; XPO1
UniProt ID: (Human) O14980, (Rat) Q80U96, (Mouse) Q6P5F9
Entrez Gene ID: (Dog) 474609, (Human) 7514, (Rat) 85252, (Mouse) 103573
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.