Proteintech
Manufacturer Code:148451AP150UL
Catalog # 50-172-9772
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf155 Cardiolipin synthase cardiolipin synthase 1 CLS CLS1 CRLS1 GCD10 Protein GCD10 homolog; cardiolipin synthase; Cardiolipin synthase (CMP-forming); cardiolipin synthetase; CLS; Protein GCD10 homolog
Gene Aliases: 0610009I22Rik; 4930557M15Rik; 5730490M08Rik; C20orf155; CLS; CLS1; CRLS1; dJ967N21.6; GCD10; RGD1311037
UniProt ID: (Human) Q9UJA2, (Mouse) Q80ZM8, (Rat) Q5U2V5
Entrez Gene ID: (Human) 54675, (Mouse) 66586, (Rat) 366196
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.