Invitrogen
Manufacturer Code:PA568023
Catalog # PIPA568023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human CRLF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: class I cytokine receptor; CLF-1; CRLM-3; cytokine receptor like molecule 3; Cytokine receptor-like factor 1; Cytokine receptor-like molecule 3; cytokine type 1 receptor CRLP-1; Cytokine-like factor 1; Novel cytokine receptor 6; NR6; UNQ288/PRO327 CISS CISS1 CLF CLF-1 NR6 zcytor5 class I cytokine receptor cytokine type 1 receptor CRLP-1 cytokine-like factor 1; ZcytoR5
Gene Aliases: CISS; CISS1; CLF; CLF-1; CRLF1; CRLM3; NR6; NR6.1; UNQ288/PRO327; zcytor5
UniProt ID: (Human) O75462, (Mouse) Q9JM58
Entrez Gene ID: (Rhesus monkey) 720247, (Human) 9244, (Mouse) 12931
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.