Abnova
Manufacturer Code:H00009244A01
Catalog # 89-106-036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 135 a.a. to 230 a.a) partial recombinant protein with GST tag.,CRLF1 (NP_004741 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant CRLF1.
For Research Use Only
Protein Aliases: CISS; CISS1; class I cytokine receptor; CLF; CLF-1; Cytokine receptor-like factor 1; cytokine type 1 receptor CRLP-1; Cytokine-like factor 1; NR6; ZcytoR5
Gene Aliases: CISS; CISS1; CLF; CLF-1; CRLF1; NR6; UNQ288/PRO327; zcytor5
UniProt ID: (Human) O75462
Entrez Gene ID: (Human) 9244
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.