R&D Systems
Manufacturer Code:MAB4675SP
Catalog # MAB4675SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 840017 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse CRISP-1 Gln20-His244 Accession # Q03401 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Acidic epididymal glycoprotein homolog; Acidic epididymal glycoprotein homolog acidic epididymal glycoprotein-like 1 AEGL1 AEG-related protein ARPAEG-like protein CRISP1 CRISP-1AEGL1HSCRISP1D cysteine-rich secretory protein 1 cysteine-rich secretory protein-1 delta HSCRISP1G HUMARP; acidic epididymal glycoprotein-like 1; AEG-like protein; AEG-related protein; ARP; CRISP-1; Cysteine-rich secretory protein 1; cysteine-rich secretory protein-1 delta; epididymis secretory protein Li 57
Gene Aliases: AEGL1; ARP; CRISP-1; CRISP1; HEL-S-57; HSCRISP1D; HSCRISP1G; HUMARP
UniProt ID: (Human) P54107
Entrez Gene ID: (Human) 167
Molecular Function:
defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.