Novus Biologicals
Manufacturer Code:NBP179533
Catalog # NBP179533
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal of human CRISP1The immunogen for this antibody is CRISP1. Peptide sequence LKMSWSEEAAQNARIFSKYCDMTESNPLERRLPNTFCGENMHMTSYPVSW. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acidic epididymal glycoprotein homolog; Acidic epididymal glycoprotein homolog acidic epididymal glycoprotein-like 1 AEG-related protein ARPAEG-like protein CRISP-1AEGL1HSCRISP1D cysteine-rich secretory protein 1 cysteine-rich secretory protein-1 delta HSCRISP1G HUMARP; acidic epididymal glycoprotein-like 1; AEG-like protein; AEG-related protein; ARP; CRISP-1; Cysteine-rich secretory protein 1; cysteine-rich secretory protein-1 delta; epididymis secretory protein Li 57
Gene Aliases: AEGL1; ARP; CRISP-1; CRISP1; HEL-S-57; HSCRISP1D; HSCRISP1G; HUMARP
UniProt ID: (Human) P54107
Entrez Gene ID: (Human) 167
Molecular Function:
defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.