Invitrogen
Manufacturer Code:PA533449
Catalog # PIPA533449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 19 amino acid peptide from N-terminal extracellular domain of human CRHR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Corticotropin-releasing factor receptor 2; Corticotropin-releasing hormone receptor 2; CRF-R-2; CRF2 receptor, beta isoform; CRH receptor 2 variant B; CRH-R-2; CRH-R2; CRHR2 CRF-R-2 CRF-R2 CRF2R Crh2 receptor CRF2 receptor beta isoform CRFR2 CRFR-2 CRH-R-2 CRH2R CRF-RB CRF2 CRH-R2 HM-CRF
Gene Aliases: CRF-RB; CRF2; CRF2R; CRFR2; CRH2R; CRHR2; HM-CRF
UniProt ID: (Human) Q13324
Entrez Gene ID: (Human) 1395
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.