Proteintech
Manufacturer Code:252671AP150UL
Catalog # 50-173-4824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: corticotrophin releasing hormone receptor 2; corticotropin releasing factor receptor type 2 beta; corticotropin-releasing factor 2; Corticotropin-releasing factor receptor 2; corticotropin-releasing factor receptor subtype 2; Corticotropin-releasing hormone receptor 2; CRF 2 receptor; CRF R 2 CRF R2 CRF2R CRFR 2 CRFR2 CRH R 2 CRH R2 CRH2R CRHR2; CRF-R 2; CRF-R-2; CRF-R2; CRF-RB; CRF2; CRF2 receptor, beta isoform; CRFR-2; CRH receptor 2 variant B; CRH-R 2; CRH-R-2; CRH-R2
Gene Aliases: CRF-R2; CRF-RB; CRF2; CRF2R; CRFR2; CRFR2alpha; CRFR2beta; CRH2R; CRHR2; HM-CRF
UniProt ID: (Human) Q13324, (Rat) P47866, (Mouse) Q60748
Entrez Gene ID: (Human) 1395, (Rat) 64680, (Mouse) 12922
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.