Abnova
Manufacturer Code:H00001395K
Catalog # 89-208-457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CRHR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CRHR2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Corticotropin-releasing factor receptor 2; Corticotropin-releasing hormone receptor 2; CRF-R-2; CRF2 receptor, beta isoform; CRFR2; CRH receptor 2 variant B; CRH-R-2; CRH-R2
Gene Aliases: CRF-RB; CRF2; CRF2R; CRFR2; CRH2R; CRHR2; HM-CRF
UniProt ID: (Human) Q13324
Entrez Gene ID: (Human) 1395
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.