R&D Systems
Manufacturer Code:FAB3930U100UG
Catalog # FAB3930U100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 343919 |
Immunogen | NS0 mouse myeloma cell line transfected with human CRHR-1 Ser24-Val415 Accession # NP_004373 |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: CHR-R1 corticotropin releasing hormone receptor 1 corticotropin-releasing factor receptor 1 corticotropin-releasing factor type 1 receptor Corticotropin-releasing hormone receptor 1 CRF1 CRFR CRF-R CRFR1 CRF-R1 CRF-R-1 CRFR-1 CRH-R-1 CRHR1f CRH-R1h CRHR1L CRHRCRH-R1 seven transmembrane helix receptor; Corticotropin-releasing factor receptor 1; corticotropin-releasing factor type 1 receptor; Corticotropin-releasing hormone receptor 1; CRF-R-1; CRH receptor 1; CRH-R-1; seven transmembrane helix receptor
Gene Aliases: CRF-R; CRF-R-1; CRF-R1; CRF1; CRFR; CRFR-1; CRFR1; CRH-R-1; CRH-R1; CRHR; CRHR1; CRHR1L
UniProt ID: (Human) P34998
Entrez Gene ID: (Human) 1394
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.